missing translation for 'onlineSavingsMsg'
Learn More

Triacetin, CP, Spectrum™ Chemical

Catalog No. p-7166651
Change view
Click to view available options
Quantity:
4 L
20 L
Packaging:
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614500 4 L Steel Can
18614499 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18614500 Supplier Spectrum Chemical Mfg Cor Supplier No. T10984LTGL
Only null left

Chemical Identifiers

CAS 102-76-1

Specifications

CAS Max % 1
Boiling Point 258°C
Packaging Steel Can
Quantity 4 L
Grade Chemically Pure
Physical Form Liquid
Chemical Name or Material Triacetin

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.