missing translation for 'onlineSavingsMsg'
Learn More

Tri-tert-butyl 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetate 97.0+%, TCI America™

Catalog No. p-7137333
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2642100MG 100 mg
T26421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2642100MG Supplier TCI America Supplier No. T2642100MG

Chemical Identifiers

CAS 137076-54-1
Molecular Formula C28H52N4O8
Molecular Weight (g/mol) 572.744
MDL Number MFCD02259697
InChI Key RVUXZXMKYMSWOM-UHFFFAOYSA-N
Synonym 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetic Acid Tri-tert-butyl Ester
PubChem CID 11606627
IUPAC Name 2-[4,7,10-tris[2-[(2-methylpropan-2-yl)oxy]-2-oxoethyl]-1,4,7,10-tetrazacyclododec-1-yl]acetic acid
SMILES CC(C)(C)OC(=O)CN1CCN(CCN(CCN(CC1)CC(=O)OC(C)(C)C)CC(=O)OC(C)(C)C)CC(=O)O

Specifications

Quantity 100 mg
Formula Weight 572.74
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tri-tert-butyl 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetraacetate
TSCA No
Recommended Storage Refrigerator