accessibility menu, dialog, popup

UserName

CH3CH3H3COONNNNOOH3CCH3H3COOCH3H3CCH3OOH

CAS RN 137076-54-1

IUPAC Name: 2-{4,7,10-tris[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetic acid
Synonyms: {4,7,10-tris[2-(tert-butoxy)-2-oxoethyl]-1,4,7,10-tetraazacyclododecan-1-yl}acetic acid
Molecular Weight (g/mol): 572.74
Molecular Formula: C28H52N4O8
InChi Key: RVUXZXMKYMSWOM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CN1CCN(CC(O)=O)CCN(CC(=O)OC(C)(C)C)CCN(CC(=O)OC(C)(C)C)CC1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results