missing translation for 'onlineSavingsMsg'
Learn More

Tri-p-tolylphosphine, 98%

Catalog No. AC422330050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422330050 5 g Glass bottle
AC422330010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422330050 Supplier Thermo Scientific Chemicals Supplier No. 422330050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1038-95-5 | C21H21P | 304.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1038-95-5
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.37
MDL Number MFCD00008542
InChI Key WXAZIUYTQHYBFW-UHFFFAOYSA-N
Synonym tri-p-tolylphosphine, tris 4-methylphenyl phosphine, tri p-tolyl phosphine, phosphine, tris 4-methylphenyl, tri-para-tolylphosphine, tris p-tolyl phosphine, phosphine, tri-p-tolyl, tris 4-methylphenyl phosphane, phosphine, tris p-tolyl, tri-p-tolyl phosphine
PubChem CID 13956
IUPAC Name tris(4-methylphenyl)phosphane
SMILES CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Melting Point 145.0°C to 150.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3C6H4)3P
Quantity 5 g
Formula Weight 304.36
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tri-p-tolylphosphine, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 213-863-5
RTECSNumber SZ3880000
TSCA TSCA

RUO – Research Use Only