missing translation for 'onlineSavingsMsg'
Learn More

Tri(p-tolyl)phosphine 96.0+%, TCI America™

Supplier:  TCI America T086225G

 View more versions of this product

Catalog No. T086225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tri(p-tolyl)phosphine
149°C
C21H21P
25 g
WXAZIUYTQHYBFW-UHFFFAOYSA-N
tris(4-methylphenyl)phosphane
13956
≥96.0% (GC)
1038-95-5
White
MFCD00008542
tri-p-tolylphosphine, tris 4-methylphenyl phosphine, tri p-tolyl phosphine, phosphine, tris 4-methylphenyl, tri-para-tolylphosphine, tris p-tolyl phosphine, phosphine, tri-p-tolyl, tris 4-methylphenyl phosphane, phosphine, tris p-tolyl, tri-p-tolyl phosphine
CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
304.37
304.37
Crystalline Powder

Chemical Identifiers

1038-95-5
304.37
WXAZIUYTQHYBFW-UHFFFAOYSA-N
13956
CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
C21H21P
MFCD00008542
tri-p-tolylphosphine, tris 4-methylphenyl phosphine, tri p-tolyl phosphine, phosphine, tris 4-methylphenyl, tri-para-tolylphosphine, tris p-tolyl phosphine, phosphine, tri-p-tolyl, tris 4-methylphenyl phosphane, phosphine, tris p-tolyl, tri-p-tolyl phosphine
tris(4-methylphenyl)phosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : SZ3880000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions