missing translation for 'onlineSavingsMsg'
Learn More

Tri(p-tolyl)phosphine 96.0+%, TCI America™

Catalog No. T08625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08625G 5 g
T086225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08625G Supplier TCI America Supplier No. T08625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1038-95-5
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.37
MDL Number MFCD00008542
InChI Key WXAZIUYTQHYBFW-UHFFFAOYSA-N
Synonym tri-p-tolylphosphine, tris 4-methylphenyl phosphine, tri p-tolyl phosphine, phosphine, tris 4-methylphenyl, tri-para-tolylphosphine, tris p-tolyl phosphine, phosphine, tri-p-tolyl, tris 4-methylphenyl phosphane, phosphine, tris p-tolyl, tri-p-tolyl phosphine
PubChem CID 13956
IUPAC Name tris(4-methylphenyl)phosphane
SMILES CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1

Specifications

Melting Point 149°C
Color White
Quantity 5 g
Formula Weight 304.37
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tri(p-tolyl)phosphine
RTECSNumber SZ3880000
TSCA Yes