missing translation for 'onlineSavingsMsg'
Learn More

Tri-o-tolylphosphine, 99%

Catalog No. AC422320250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422320250 25 g Glass bottle
AC422320010 1 g Glass bottle
AC422320050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC422320250 Supplier Thermo Scientific Chemicals Supplier No. 422320250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6163-58-2 | C21H21P | 304.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6163-58-2
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.37
MDL Number MFCD00008514
InChI Key COIOYMYWGDAQPM-UHFFFAOYSA-N
Synonym tri-o-tolylphosphine, tri o-tolyl phosphine, tris 2-methylphenyl phosphine, tris 2-methylphenyl phosphane, phosphine, tris 2-methylphenyl, tris o-tolyl phosphine, tri-ortho-toylphosphine, tri-ortho-tolylphosphine, phosphine, tri-o-tolyl, tri 2-methylphenyl phosphine
PubChem CID 80271
IUPAC Name tris(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C

Specifications

Melting Point 126.0°C to 127.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3C6H4)3P
Quantity 25 g
Formula Weight 304.37
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tri-o-tolylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 228-193-9

RUO – Research Use Only