missing translation for 'onlineSavingsMsg'
Learn More

Tri-o-tolylbismuth Dichloride 97.0+%, TCI America™

$346.71 - $1,193.00

Chemical Identifiers

CAS 6729-60-8
Molecular Formula C21H21BiCl2
Molecular Weight (g/mol) 553.279
MDL Number MFCD00092459
InChI Key RLWWKAGRZATJDC-UHFFFAOYSA-L
Synonym Dichloro(tri-o-tolyl)bismuth, Tris(2-methylphenyl)bismuth Dichloride
PubChem CID 5225331
IUPAC Name dichloro-tris(2-methylphenyl)bismuth
SMILES CC1=CC=CC=C1[Bi](C2=CC=CC=C2C)(C3=CC=CC=C3C)(Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T19571G
View Documents
TCI America
T19571G
1 g N/A
Only null left
 
T19575G
View Documents
TCI America
T19575G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6729-60-8
553.279
RLWWKAGRZATJDC-UHFFFAOYSA-L
5225331
CC1=CC=CC=C1[Bi](C2=CC=CC=C2C)(C3=CC=CC=C3C)(Cl)Cl
C21H21BiCl2
MFCD00092459
Dichloro(tri-o-tolyl)bismuth, Tris(2-methylphenyl)bismuth Dichloride
dichloro-tris(2-methylphenyl)bismuth

Specifications

6729-60-8
MFCD00092459
Dichloro(tri-o-tolyl)bismuth, Tris(2-methylphenyl)bismuth Dichloride
CC1=CC=CC=C1[Bi](C2=CC=CC=C2C)(C3=CC=CC=C3C)(Cl)Cl
553.279
553.28
Crystalline Powder
C21H21BiCl2
1 g
RLWWKAGRZATJDC-UHFFFAOYSA-L
dichloro-tris(2-methylphenyl)bismuth
5225331
≥97.0% (T)
Tri-o-tolylbismuth Dichloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'