accessibility menu, dialog, popup

UserName

CAS RN 6729-60-8

IUPAC Name: dichlorotris(2-methylphenyl)bismuthane
Synonyms: dichlorotris(2-methylphenyl)bismuthane
Molecular Weight (g/mol): 553.28
Molecular Formula: C21H21BiCl2
InChi Key: RLWWKAGRZATJDC-UHFFFAOYSA-L
SMILES: CC1=CC=CC=C1[Bi](Cl)(Cl)(C1=CC=CC=C1C)C1=CC=CC=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results