missing translation for 'onlineSavingsMsg'
Learn More

Tri-o-tolylbismuth Dichloride 97.0+%, TCI America™

Catalog No. T19571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19571G 1 g
T19575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19571G Supplier TCI America Supplier No. T19571G

Chemical Identifiers

CAS 6729-60-8
Molecular Formula C21H21BiCl2
Molecular Weight (g/mol) 553.279
MDL Number MFCD00092459
InChI Key RLWWKAGRZATJDC-UHFFFAOYSA-L
Synonym Dichloro(tri-o-tolyl)bismuth, Tris(2-methylphenyl)bismuth Dichloride
PubChem CID 5225331
IUPAC Name dichloro-tris(2-methylphenyl)bismuth
SMILES CC1=CC=CC=C1[Bi](C2=CC=CC=C2C)(C3=CC=CC=C3C)(Cl)Cl

Specifications

Quantity 1 g
Formula Weight 553.28
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tri-o-tolylbismuth Dichloride
TSCA No
Recommended Storage Refrigerator