missing translation for 'onlineSavingsMsg'
Learn More

Tri(o-tolyl)phosphine 97.0+%, TCI America™

Catalog No. T10245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10245G 5 g
T102425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10245G Supplier TCI America Supplier No. T10245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6163-58-2
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.373
MDL Number MFCD00008514
InChI Key COIOYMYWGDAQPM-UHFFFAOYSA-N
Synonym tri-o-tolylphosphine, tri o-tolyl phosphine, tris 2-methylphenyl phosphine, tris 2-methylphenyl phosphane, phosphine, tris 2-methylphenyl, tris o-tolyl phosphine, tri-ortho-toylphosphine, tri-ortho-tolylphosphine, phosphine, tri-o-tolyl, tri 2-methylphenyl phosphine
PubChem CID 80271
IUPAC Name tris(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 5 g
Formula Weight 304.37
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tri(o-tolyl)phosphine
EINECSNumber (3)-4357
TSCA Yes