missing translation for 'onlineSavingsMsg'
Learn More

Tri-o-cresyl Phosphate 97.0+%, TCI America™

Supplier:  TCI America P13315G

 View more versions of this product

Catalog No. P13315G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tri-o-cresyl Phosphate
Yellow
C21H21O4P
5 g
tri-o-cresyl phosphate, tri-o-tolyl phosphate, o-tolyl phosphate, tocp, totp, phosphoric acid, tris 2-methylphenyl ester, o-cresyl phosphate, triorthocresyl phosphate, phosflex 179c, tri-2-tolyl phosphate
CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C
368.37
368.37
Liquid
78-30-8
410°C
MFCD00041906
2574
YSMRWXYRXBRSND-UHFFFAOYSA-N
tris(2-methylphenyl) phosphate
6527
≥97.0% (GC)

Chemical Identifiers

78-30-8
368.37
YSMRWXYRXBRSND-UHFFFAOYSA-N
6527
CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C
C21H21O4P
MFCD00041906
tri-o-cresyl phosphate, tri-o-tolyl phosphate, o-tolyl phosphate, tocp, totp, phosphoric acid, tris 2-methylphenyl ester, o-cresyl phosphate, triorthocresyl phosphate, phosflex 179c, tri-2-tolyl phosphate
tris(2-methylphenyl) phosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2522&(3)-3363&(3)-2613

RTECSNumber : TD0350000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions