missing translation for 'onlineSavingsMsg'
Learn More

Tri-o-cresyl Phosphate 97.0+%, TCI America™

Catalog No. p-7135081
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13315G 5 g
P133125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13315G Supplier TCI America Supplier No. P13315G

Chemical Identifiers

CAS 78-30-8
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.37
MDL Number MFCD00041906
InChI Key YSMRWXYRXBRSND-UHFFFAOYSA-N
Synonym tri-o-cresyl phosphate, tri-o-tolyl phosphate, o-tolyl phosphate, tocp, totp, phosphoric acid, tris 2-methylphenyl ester, o-cresyl phosphate, triorthocresyl phosphate, phosflex 179c, tri-2-tolyl phosphate
PubChem CID 6527
IUPAC Name tris(2-methylphenyl) phosphate
SMILES CC1=CC=CC=C1OP(=O)(OC1=CC=CC=C1C)OC1=CC=CC=C1C

Specifications

Color Yellow
Boiling Point 410°C
Quantity 5 g
UN Number 2574
Formula Weight 368.37
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tri-o-cresyl Phosphate
EINECSNumber (3)-2522&(3)-3363&(3)-2613
RTECSNumber TD0350000
TSCA Yes