missing translation for 'onlineSavingsMsg'
Learn More

Tri-O-benzyl-D-glucal 95.0+%, TCI America™

Catalog No. T18591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18591G 1 g
T18595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18591G Supplier TCI America Supplier No. T18591G

Chemical Identifiers

CAS 55628-54-1
Molecular Formula C27H28O4
Molecular Weight (g/mol) 416.517
MDL Number MFCD00061640
InChI Key MXYLLYBWXIUMIT-PFBJBMPXSA-N
Synonym 3,4,6-tri-o-benzyl-d-glucal, tri-o-benzyl-d-glucal, 2r,3s,4r-3,4-bis benzyloxy-2-benzyloxy methyl-3,4-dihydro-2h-pyran, pubchem10548, 1,5-anhydro-3,4,6-tri-o-benzyl-2-deoxy-d-arabinohex-1-enitol, 1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol 3,4,6-tribenzyl ether
PubChem CID 11742644
IUPAC Name (2R,3S,4R)-3,4-bis(phenylmethoxy)-2-(phenylmethoxymethyl)-3,4-dihydro-2H-pyran
SMILES C1=CC=C(C=C1)COCC2C(C(C=CO2)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 1 g
Formula Weight 416.52
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tri-O-benzyl-D-glucal
TSCA No
Recommended Storage Refrigerator