missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-propyl orthoformate, 97%

Catalog No. p-7022633
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1550306 5 g
AAA1550314 25 g
AAA1550322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1550306 Supplier Thermo Scientific Chemicals Supplier No. A1550306

CAS: 621-76-1 | C10H22O3 | 190.283 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-76-1
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.283
MDL Number MFCD00015214
InChI Key RWNXXQFJBALKAX-UHFFFAOYSA-N
Synonym tripropyl orthoformate, tri-n-propyl orthoformate, orthoformic acid tripropyl ester, 1-dipropoxymethoxy propane, unii-z8f0df6af9, propane, 1,1',1-methylidynetris oxy tris, z8f0df6af9, tripropoxymethane, tripropylorthoformate
PubChem CID 69311
IUPAC Name 1-(dipropoxymethoxy)propane
SMILES CCCOC(OCCC)OCCC

Specifications

Density 0.886
Boiling Point 106°C to 108°C (40 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.407
Quantity 5 g
Beilstein 1744249
Sensitivity Moisture sensitive
Formula Weight 190.28
Percent Purity 97%
Chemical Name or Material Tri-n-propyl orthoformate
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 210-704-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only