missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylamine 97.0+%, TCI America™

Supplier:  TCI America T0502100ML

 View more versions of this product

Catalog No. T0502100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tri-n-octylamine
Yellow
C24H51N
100 mL
XTAZYLNFDRKIHJ-UHFFFAOYSA-N
N,N-dioctyloctan-1-amine
14227
≥97.0% (GC,T)
1116-76-3
367°C
MFCD00009560
trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
CCCCCCCCN(CCCCCCCC)CCCCCCCC
353.679
353.68
Liquid

Chemical Identifiers

1116-76-3
353.679
XTAZYLNFDRKIHJ-UHFFFAOYSA-N
14227
CCCCCCCCN(CCCCCCCC)CCCCCCCC
C24H51N
MFCD00009560
trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
N,N-dioctyloctan-1-amine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0143&(2)-0139

RTECSNumber : RG8225000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions