missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylamine 97.0+%, TCI America™

Catalog No. p-7136313
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T050225ML 25 mL
T0502100ML 100 mL
T0502500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T050225ML Supplier TCI America Supplier No. T050225ML
Only null left

Chemical Identifiers

CAS 1116-76-3
Molecular Formula C24H51N
Molecular Weight (g/mol) 353.679
MDL Number MFCD00009560
InChI Key XTAZYLNFDRKIHJ-UHFFFAOYSA-N
Synonym trioctylamine, tri-n-octylamine, tricaprylamine, tricaprylylamine, 1-octanamine, n,n-dioctyl, tri-n-caprylylamine, alamine 336, alamine 336s, alamine 3365, farmin 08
PubChem CID 14227
IUPAC Name N,N-dioctyloctan-1-amine
SMILES CCCCCCCCN(CCCCCCCC)CCCCCCCC

Specifications

Color Yellow
Boiling Point 367°C
Quantity 25 mL
Formula Weight 353.68
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Tri-n-octylamine
EINECSNumber (2)-0143&(2)-0139
RTECSNumber RG8225000
TSCA Yes