missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-decylamine 97.0+%, TCI America™

Catalog No. p-7136270
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04185G 5 g
T041825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04185G Supplier TCI America Supplier No. T04185G
Only null left

Chemical Identifiers

CAS 1070-01-5
Molecular Formula C30H63N
Molecular Weight (g/mol) 437.841
MDL Number MFCD00041914
InChI Key COFKFSSWMQHKMD-UHFFFAOYSA-N
Synonym tri-n-decylamine, tris decyl amine, 1-decanamine, n,n-didecyl, tridecanitrile mixed isomers, trisdecylamine, tri decyl amine, n,n-didecyl-1-decanamine, 4-04-00-00785 beilstein handbook reference, tri-n-decylamine tertiary amine
PubChem CID 14049
IUPAC Name N,N-didecyldecan-1-amine
SMILES CCCCCCCCCCN(CCCCCCCCCC)CCCCCCCCCC

Specifications

Color Yellow
Boiling Point 430°C
Quantity 5 g
Formula Weight 437.84
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Tri-n-decylamine
EINECSNumber (2)-0139
RTECSNumber YD3675000
TSCA Yes