accessibility menu, dialog, popup

UserName

CH3NCH3CH3

CAS RN 1070-01-5

IUPAC Name: tris(decyl)amine
Synonyms: 1-decanamine, N,N-didecyl-
Molecular Weight (g/mol): 437.84
Molecular Formula: C30H63N
InChi Key: COFKFSSWMQHKMD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCN(CCCCCCCCCC)CCCCCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results