missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butylamine, 98%

Catalog No. aaa148680f
Change view
Click to view available options
Quantity:
10,000 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA148680F 2500 mL
AAA14868AP 500 mL
AAA148680D 10,000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA148680F Supplier Thermo Scientific Chemicals Supplier No. A148680F
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 102-82-9 | C12H27N | 185.355 g/mol

Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.

Solubility
Miscible with acetone, ethanol, ethyl ether and most organic solvents. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 102-82-9
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.355
MDL Number MFCD00009431
InChI Key IMFACGCPASFAPR-UHFFFAOYSA-N
Synonym tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7
PubChem CID 7622
ChEBI CHEBI:38905
IUPAC Name N,N-dibutylbutan-1-amine
SMILES CCCCN(CCCC)CCCC

Specifications

Melting Point <-70°C
Density 0.778
pH 10.6
Boiling Point 216°C
Flash Point 71°C (159°F)
Odor Amine-like
Refractive Index 1.429
Linear Formula [CH3(CH2)3]3N
Quantity 2500 mL
UN Number UN2542
Beilstein 1698872
Merck Index 14,9618
Solubility Information Miscible with acetone,ethanol,ethyl ether and most organic solvents. Immiscible with water.
Formula Weight 185.36
Percent Purity 98%
Chemical Name or Material Tri-n-butylamine
Show More Show Less
Hazard Category H227-H302-H310+H330-H315-H319
Hazard Statement GHS H Statement
H310-H330-H227-H302-H315-H319
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P235-P260-P262-P264b-P270-P271-P280-P284-P301+P312-P302+P350-P304+P340-P305+P351+P338-P310-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TRIBUTYLAMINE
EINECSNumber 203-058-7
RTECSNumber YA0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only