Learn More
Tri-n-butylamine, 98%
Description
Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.
Solubility
Miscible with acetone, ethanol, ethyl ether and most organic solvents. Immiscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | <-70°C |
| Density | 0.778 |
| pH | 10.6 |
| Boiling Point | 216°C |
| Flash Point | 71°C (159°F) |
| Odor | Amine-like |
| Refractive Index | 1.429 |
| Linear Formula | [CH3(CH2)3]3N |
| Quantity | 10,000 mL |
| UN Number | UN2542 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302-H310+H330-H315-H319 |
| Hazard Statement | GHS H Statement H310-H330-H227-H302-H315-H319 Fatal in contact with skin. Fatal if inhaled. Combustible liquid. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P210-P235-P260-P262-P264b-P270-P271-P280-P284-P301+P312-P302+P350-P304+P340-P305+P351+P338-P310-P330-P332+P313-P361-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TRIBUTYLAMINE |
| EINECSNumber | 203-058-7 |
| RTECSNumber | YA0350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only