missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals

Catalog No. AAA16084AK
Change view
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA16084AK 250 mL
AAA16084AU 1000 mL
AAA160840J 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA16084AK Supplier Thermo Scientific Chemicals Supplier No. A16084AK

Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.

Solubility
Miscible with diethyl ether, benzene, carbon disulfide and ethanol. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and alkali.

Chemical Identifiers

CAS 126-73-8
Molecular Formula C12H27O4P
Molecular Weight (g/mol) 266.32
MDL Number MFCD00009436
InChI Key STCOOQWBFONSKY-UHFFFAOYSA-N
Synonym tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de
PubChem CID 31357
ChEBI CHEBI:35019
IUPAC Name tributyl phosphate
SMILES CCCCOP(=O)(OCCCC)OCCCC

Specifications

Melting Point -80°C
Density 0.977
Boiling Point 155°C to 156°C (10 mmHg)
Flash Point 152°C (305°F)
Odor Odorless
Assay Percent Range 98%
Refractive Index 1.424
Quantity 250 mL
Beilstein 1710584
Merck Index 14,9618
Solubility Information Miscible with diethyl ether,benzene,carbon disulfide and ethanol. Slightly miscible with water.
Formula Weight 266.32
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tri-n-butyl phosphate
Show More Show Less
Hazard Category H302-H315-H319-H351
Hazard Statement GHS H Statement
H351-H302-H315
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement P201-P202-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 204-800-2
RTECSNumber TC7700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only