missing translation for 'onlineSavingsMsg'
Learn More

Tri(m-tolyl)phosphine 98.0+%, TCI America™

Catalog No. p-7136558
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10255G 5 g
T102525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10255G Supplier TCI America Supplier No. T10255G

Chemical Identifiers

CAS 6224-63-1
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.373
MDL Number MFCD00008532
InChI Key LFNXCUNDYSYVJY-UHFFFAOYSA-N
Synonym tri-m-tolylphosphine, tris 3-methylphenyl phosphine, tri m-tolyl phosphine, phosphine, tris 3-methylphenyl, tris m-tolyl phosphine, phosphine, tri-m-tolyl, tris 3-methylphenyl phosphane, tris 3-tolyl phosphine, tri-m-tolyl-phosphane, tri-m-tolylphosphane
PubChem CID 80362
IUPAC Name tris(3-methylphenyl)phosphane
SMILES CC1=CC(=CC=C1)P(C2=CC=CC(=C2)C)C3=CC=CC(=C3)C

Specifications

Melting Point 100°C
Color White-Yellow
Quantity 5 g
Formula Weight 304.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tri(m-tolyl)phosphine
EINECSNumber (3)-3479
TSCA Yes