missing translation for 'onlineSavingsMsg'
Learn More

Tri-m-cresyl Phosphate 95.0+%, TCI America™

Catalog No. P14725G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P14725G Supplier TCI America Supplier No. P14725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 563-04-2
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.37
MDL Number MFCD00041907
InChI Key RMLPZKRPSQVRAB-UHFFFAOYSA-N
Synonym tri-m-cresyl phosphate, tri-m-tolyl phosphate, phosphoric acid, tris 3-methylphenyl ester, tri-m-cresyl phosphite, tris-m-cresyl phosphate, tris m-tolyl phosphate, tris 3-methylphenyl phosphate, phosphoric acid tri-m-tolyl ester, unii-w09q34on6j, ccris 5948
PubChem CID 11232
IUPAC Name tris(3-methylphenyl) phosphate
SMILES CC1=CC(OP(=O)(OC2=CC(C)=CC=C2)OC2=CC=CC(C)=C2)=CC=C1

Specifications

Color White-Yellow
Boiling Point 240°C
Quantity 5 g
UN Number 3077
Formula Weight 368.37
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Tri-m-cresyl Phosphate
EINECSNumber (3)-2522&(3)-2613&(3)-3363
TSCA Yes