accessibility menu, dialog, popup

UserName

CAS RN 563-04-2

IUPAC Name: tris(3-methylphenyl) phosphate
Synonyms: tri-M-cresyl phosphate
Molecular Weight (g/mol): 368.37
Molecular Formula: C21H21O4P
InChi Key: RMLPZKRPSQVRAB-UHFFFAOYSA-N
SMILES: CC1=CC(OP(=O)(OC2=CC(C)=CC=C2)OC2=CC=CC(C)=C2)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results