missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri(2-thienyl)phosphine 96.0+%, TCI America™
Supplier: TCI America T16665G
Specifications
| Tri(2-thienyl)phosphine | |
| 205°C | |
| MFCD00015471 | |
| tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine # | |
| S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1 | |
| 280.36 | |
| 280.36 | |
| Crystalline Lumps |
| 24171-89-9 | |
| C12H9PS3 | |
| 5 g | |
| KUCPTMZJPDVWJL-UHFFFAOYSA-N | |
| tris(thiophen-2-yl)phosphane | |
| 90384 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 24171-89-9 | |
| 280.36 | |
| KUCPTMZJPDVWJL-UHFFFAOYSA-N | |
| 90384 | |
| S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1 |
| C12H9PS3 | |
| MFCD00015471 | |
| tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine # | |
| tris(thiophen-2-yl)phosphane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator