missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri(2-thienyl)phosphine 96.0+%, TCI America™
$270.29 - $1002.86
Chemical Identifiers
| CAS | 24171-89-9 |
|---|---|
| Molecular Formula | C12H9PS3 |
| Molecular Weight (g/mol) | 280.36 |
| MDL Number | MFCD00015471 |
| InChI Key | KUCPTMZJPDVWJL-UHFFFAOYSA-N |
| Synonym | tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine # |
| PubChem CID | 90384 |
| IUPAC Name | tris(thiophen-2-yl)phosphane |
| SMILES | S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1 |
Chemical Identifiers
| 24171-89-9 | |
| 280.36 | |
| KUCPTMZJPDVWJL-UHFFFAOYSA-N | |
| 90384 | |
| S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1 |
| C12H9PS3 | |
| MFCD00015471 | |
| tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine # | |
| tris(thiophen-2-yl)phosphane |
Specifications
| 24171-89-9 | |
| C12H9PS3 | |
| 1 g | |
| KUCPTMZJPDVWJL-UHFFFAOYSA-N | |
| tris(thiophen-2-yl)phosphane | |
| 90384 | |
| ≥96.0% (GC) | |
| Tri(2-thienyl)phosphine |
| 205°C | |
| MFCD00015471 | |
| tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine # | |
| S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1 | |
| 280.36 | |
| 280.36 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator