missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri(2-furyl)phosphine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 63°C |
| Boiling Point | 136°C |
| Quantity | 1 g |
| Formula Weight | 232.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tri(2-furyl)phosphine |
Safety and Handling
| TSCA | No |