missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Trenbolone Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA175009
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA175009 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA175009 Supplier Invitrogen™ Supplier No. PA175009

Sheep Polyclonal Antibody

Working Concentration: 0.25 μg/mL. Sensitivity: 10 ng/mL Alpha trenbolone produces 93% inhibition in a competitive ELISA employing 17αTrenbolone polyclonal antibody.

Specifications

Antigen Trenbolone
Applications ELISA
Classification Polyclonal
Concentration 9.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Alias 17-beta-Trenbolone; 17beta-Trenbolone; C18H22O2; Trenbolonum; Trienbolone
Host Species Sheep
Immunogen 17a-Trenbolone(3)-BTG.
Purification Method Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less