missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Travoprost Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 157283-68-6 |
| Linear Formula | C26H35F3O6 |
| Synonym | Travoprost solution |
| Type | Certified Reference Material |
| Quantity | 3 mL |
| Formula Weight | 500.55 |
| Grade | Certified Reference Material |