missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Trap-Pak™ Bags (Medium)

Catalog No. GEN084033
Encompass
Change view
Click to view available options
Quantity:
1 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GEN084033 1 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GEN084033 Supplier Applied Biosystems™ Supplier No. GEN084033

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Trap-Pak™ bags are tightly sealed membrane packets containing molecular sieves to reduce and maintain a low water content in solutions that must remain anhydrous.

Used With:Anhydrous Acetonitrile, Amidite Diluent, Tetrazole/Acetonitrile, Amidite Activator Solution, Tetrazole/Acetonitrile, Amidite Activator Solution, Tetrazole/Acetonitrile, Amidite Activator Solution

Specifications

Content And Storage This product may be stored at room temperature.
Sample Type DNA
For Use With (Equipment) 3900 DNA Synthesizer
For Use With (Application) Oligo Synthesis
Product Line Trap-Pak
Product Type Bag
Quantity 1 Pk.
Reagent Type Solvent or General Reagent
Shipping Condition Room Temperature
Form Solid

For Research Use Only. Not for use in diagnostic procedures.