missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Translucent Snap Caps for 12 mm Tubes

Catalog No. p-9726002
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Natural
Red
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22010020 Gray
22010021 Blue
22010022 Natural
22010023 Green
22010024 Lavender
22010025 Red
22010026 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22010020 Supplier Globe Scientific Supplier No. 113142A
Only null left

These colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes, and plastic culture tubes.

These colorful snap-on closures are precision molded from low-density polyethylene (LDPE) and maintain a tight seal during centrifugation, agitation and refrigeration procedures. They are available in bright translucent colors for sample identification purposes. The caps feature a single thumb tab for easy removal and close nesting in tube racks.

  • Economical way to recap glass tubes, regardless of manufacturer
  • Available in seven transparent colors that correspond to Vacutainer™ cap colors: Natural, Blue, Green, Lavender, Red, Gray, and Yellow
  • Caps can be written on with indelible ink for easy sample identification
  • Prevents spills and evaporation by employing a liquid tight seal

Specifications

Product Type Snap Cap
Closure Size 12/13 mm
Autoclavable Not Autoclavable
Material LDPE
Color Gray
No. per Pack 1000
For Use With (Equipment) 12 mm Glass & Plastic Tubes / 13 mm Evacuated Tubes
Disposable Single-use