missing translation for 'onlineSavingsMsg'
Learn More

Transformer oil (PCB free) , MilliporeSigma™ Supelco™

Catalog No. 111001616
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111001616 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001616 Supplier Merck Emd Millipore Supplier No. 40900U
Only null left
Environmental

Specifications

CAS 64742-46-7
Flash Point 146°C
Type Organic Pollutant Standard
Quantity 250 mL
Grade Analytical Standard