Learn More
Invitrogen™ Transferrin From Human Serum, Alexa Fluor™ Conjugate
Description
Fluorescent transferrin for receptor-mediated endocytosis and recycling
Fluorescent transferrin conjugates are widely used to study transferrin receptor-mediated endocytosis, endosomal trafficking, and recycling using live-cell imaging or fixed-cell endpoint analysis. Transferrin binds transferrin receptor 1 (TfR1) at the cell surface and is internalized through clathrin-mediated endocytosis. Following endosomal acidification and iron release, apo-transferrin remains bound to TfR1 and the complex recycles to the plasma membrane, where apo-transferrin dissociates.
Why use fluorescent transferrin for endocytosis studies?
- Track receptor-mediated endocytosis: Follow transferrin binding, TfR-mediated internalization, and intracellular trafficking.
- Monitor endocytic recycling: Use fluorescent transferrin as a well-established marker of the endosomal recycling pathway.
- Study trafficking kinetics: Follow uptake and recycling using live-cell imaging, pulse-chase experiments, or fixed-cell endpoint analysis.
- Assess endosomal trafficking: Investigate trafficking events associated with endosomal acidification, cargo sorting, and membrane transport.
- Compare trafficking pathways: Combine fluorescent transferrin with fluorescent LDL or other cargo markers to compare recycling and lysosomally directed pathways.
- Enable multiplex fluorescence imaging: Choose from fluorescein and Alexa Fluor™ 488, 546, 555, 568, 594, 633, and 647 conjugates for multicolor imaging workflows.
Fluorescent transferrin enables a direct fluorescence readout of a well-characterized endocytic pathway, making it a versatile probe for studying receptor internalization, endosomal trafficking, and recycling.
Choose a fluorescent transferrin conjugate for your imaging workflow
Select from fluorescein and Alexa Fluor™ 488, 546, 555, 568, 594, 633, and 647 transferrin conjugates, with fluorescence spanning green to far-red wavelengths.
Multiple fluorophore options
- Match your imaging system: Select a conjugate compatible with available excitation sources, emission filters, and detectors.
- Support multicolor imaging: Choose spectrally distinct labels for use with antibodies, organelle markers, and other fluorescent probes.
- Optimize experimental design: Select the excitation and emission profile that best fits your available fluorescence channels.
- Use one biological probe across workflows: Study transferrin receptor trafficking with fluorophore options suitable for a range of fluorescence microscopy applications.
Applications of fluorescent transferrin
Fluorescent transferrin is a well-established probe for studying transferrin receptor-mediated endocytosis and intracellular trafficking.
Use fluorescent transferrin to:
- Visualize receptor-mediated internalization and recycling using live-cell imaging or fixed-cell endpoint analysis
- Track endosomal trafficking using fluorescence and confocal microscopy
- Study trafficking events associated with endosomal acidification and changes in endosomal function
- Investigate transferrin receptor dynamics, including appropriately designed FRET-based studies
- Measure transferrin receptor binding in mammalian and parasite model systems
- Compare recycling and lysosomal trafficking using complementary fluorescent cargo markers
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | Store in freezer (-5°C to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| Product Type | Transferrin |
| Dye Type | Alexa Fluor Dyes |
| Form | Solid |
| pH | 7.2 |
| Product Line | Alexa Fluor |
| Quantity | 5 mg |
| Shipping Condition | Room Temperature |
| Excitation/Emission | 495/519 nm |
| Show More |
Frequently Asked Questions (FAQs)
Transferrin From Human Serum, Alexa Fluor 568 Conjugate is holotransferrin (saturated with iron). Lysine residues of transferrin are used for conjugation of the fluorophore. On the Certificate of Analysis, you can find information regarding the degree of labeling (number of fluorophores per one protein molecule).
For Research Use Only. Not for use in diagnostic procedures.