missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Transferpette™ Single Channel Electronic Pipet PROMO

Catalog No. 13668151TU
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13668151TU 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13668151TU Supplier BRANDTECH™ BRAND™ Supplier No. BRT705320TU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.

  • Transferpette electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes with the advantages of an electronic device
  • The pipette shaft can be unscrewed and is entirely autoclavable (121°C).
  • Five convenient programs: pipetting, reverse pipetting, mixing, GEL-Electrophoresis and dispensing.
  • Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools.
  • Limitations of use: Operating temperature of instrument and reagent should be between +15°C and +40°C (59°F to 104°F) (other temperatures upon request)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s
  • For viscous media, the speed must be adjusted if necessary.

Specifications

Accuracy ≤ ± 1.0%, 0.2μL
Battery Type Rechargeable NiMH
Certifications/Compliance DIN EN ISO 8655-2
Autoclavable Autoclavable
Color Gray
Number of Channels 1
Product Line Transferpette
Type Single Channel Microliter Pipette
Volume (Metric) 2 to 20 μL
Display LCD
Finger Style Ergonomic
Includes USA/Japan AC Adapter
Increments 0.02 μL
Quantity 1 Ea.
Temperature (English) Operating 59°F to 104°F
Temperature (Metric) Operating 15°C to 40°C
Warranty 1 Year
Electrical Requirements 110V/50-60 Hz
Frequency 50/60 Hz
Show More Show Less