missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ Transferpette™ -8/-12 electronic Multichannel Pipette - TRADE-UP PROMO
Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.
Supplier: BRANDTECH™ BRAND™ 705474TUDUP
Description
- Choice of 8 or 12 channel models
- Clear, legible LCD screen
- Intuitive interface with user-friendly desing allows easy operation
- Soft touch pipetting key and adjustable finger rest provides added comfort
- Rechargeable battery provides over 4000 pipetting cycles between charges
- Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
- Easily maintained
Operating modes
- Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
- Mixing of samples (PIPmix Mode): program for mixing liquids
- Reverse Pipetting (revPIP Mode): program designed for greater accuracy
- Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
- Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots
Specifications
| ±1.6μL | |
| 2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip, 5 to 200μL tip | |
| Autoclavable | |
| Reusable | |
| Ergonomic | |
| Transferpette -8/-12 Electronic | |
| Transferpette | |
| Non-sterile | |
| 10 to 200 μL | |
| Ergonomic | |
| 0.2μL | |
| 0.25% (0.5 μL) |
| Rechargeable | |
| No | |
| Gray | |
| Pipette Tip (200 μL) | |
| Plastic | |
| 12 | |
| Step-based | |
| Electronic Pipette | |
| LCD | |
| Battery Pack, Universal Power Adapter with Charging Cable, Shelf Mount, Silicone O-rings, Reagent Reservoir, Grease, Operating Manual | |
| Yes | |
| 1 Year |