missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Transferpette™ -8/-12 electronic Multichannel Pipette - TRADE-UP PROMO

Combines comfortable design, balanced weight distribution, intuitive software and user-friendly documentation for your pipetting needs.

Supplier:  BRANDTECH™ BRAND™ 705423TU

 View more versions of this product

Catalog No. 14389125TU

Only null left
Explore available promotions

Description

Description

  • Choice of 8 or 12 channel models
  • Clear, legible LCD screen
  • Intuitive interface with user-friendly desing allows easy operation
  • Soft touch pipetting key and adjustable finger rest provides added comfort
  • Rechargeable battery provides over 4000 pipetting cycles between charges
  • Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
  • Easily maintained

Operating modes

  • Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
  • Mixing of samples (PIPmix Mode): program for mixing liquids
  • Reverse Pipetting (revPIP Mode): program designed for greater accuracy
  • Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
  • Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots
Specifications

Specifications

±0.8μL
2 to 200μL tip, 5 to 300μL tip, 2 to 20μL tip, 5 to 100μL tip, 5 to 200μL tip
Autoclavable
Reusable
Ergonomic
Transferpette -8/-12 Electronic
Transferpette
Non-sterile
5 to 100 μL
Ergonomic
0.2μL
0.25% (0.25 μL)
Rechargeable
No
Gray
Pipette Tip (200 μL)
Plastic
8
Step-based
Electronic Pipette
LCD
Battery Pack, Universal Power Adapter with Charging Cable, Shelf Mount, Silicone O-rings, Reagent Reservoir, Grease, Operating Manual
Yes
1 Year
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions