Learn More
BrandTech™ Transferpette™ -8/-12 electronic Multichannel Pipette TRADE IN PROMO
Versatile pipette offers five operating modes to improve productivity and increase sample throughput
Supplier:  BrandTech™ 705473TU
Catalog No. 13668427TU
Only null left
Description
Description
  • Choice of 8 or 12 channel models
  • Clear, legible LCD screen
  • Intuitive interface with user-friendly desing allows easy operation
  • Soft touch pipetting key and adjustable finger rest provides added comfort
  • Rechargeable battery provides over 4000 pipetting cycles between charges
  • Multichannel V-rings on tip cones improves tip fit and reduces mounting and ejection force
  • Easily maintained
Operating modes
  • Pipetting (PIP Mode): previously set volume is aspirated into pipette tip and then discharged
  • Mixing of samples (PIPmix Mode): program for mixing liquids
  • Reverse Pipetting (revPIP Mode): program designed for greater accuracy
  • Pipetting for electrophoresis (GEL Mode): program for loading of electrophoresis gels
  • Dispensing (DISP Mode): single aspirated volume is dispensed in selected number of smaller, equal aliquots
Specifications
Specifications
±0.8 μL
2 to 200 μL tip, 5 to 300 μL tip, 2 to 20 μL tip, 5 to 100 μL tip, 5 to 200 μL tip
12
Step-based
5 to 100 μL
Ergonomic
0.2 μL
1 Year
Rechargeable
Transferpette™ -8/-12 Electronic
Transferpette
Electronic Pipette
LCD
Battery, power supply unit, 120VAC adapter with charging cable, operating manual, pipette stand, reagent reservoir and five operating modes
Yes
Product Suggestions
Product Suggestions