missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ Transferpette™ -8/-12 Electronic Multichannel Pipette PROMO

Catalog No. 13668422TU
Change view
Click to view available options
Number of Channels:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
13668422TU 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13668422TU Supplier BRANDTECH™ Supplier No. BRT705419TU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Versatile pipette offering five operating modes to improve productivity and increase sample throughput.

  • Transferpette™ electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes with the advantages of an electronic device.
  • Features: The pipette shaft can be unscrewed and is entirely autoclavable (121°C).
  • Functions: pipetting, reverse pipetting, mixing, dispensing, up to 300 μL also GEL-Electrophoresis.
  • Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools.
  • Limitations of use: Operating temperature of instrument and reagent should be between +15°C and +40°C (59 °F to 104 °F) (other temperatures upon request)
  • Vapor pressure up to 500 mbar
  • Viscosity: 260 mPa s
  • For viscous media, the speed must be adjusted if necessary.

Specifications

Accuracy ±0.12 μL
Battery Type Rechargeable
Compatible Tips 0.1 to 20 μL Nano-cap Tip, 0.5 to 20 μL Ultra Micro Tip, 0.1 to 1 μL Nano-cap Tip, 0.5 to 10 μL Tip
Model DE-M
Number of Channels 8
Product Line Transferpette
Programmability Step-based
Type Electronic Pipette
Volume (Metric) 0.5 to 10 μL
Display LCD
Finger Style Ergonomic
Includes Battery, Power Supply Unit, 120 VAC Adapter with Charging Cable, Operating Manual, Pipette Stand, Reagent Reservoir
Increments 0.02 μL
Multilingual User Interface Yes
Warranty 1 Year
Show More Show Less