missing translation for 'onlineSavingsMsg'
Learn More

Poltex Transfer Pipette Cup

Catalog No. 22000424
Encompass
Change view
Click to view available options
Mounting:
Powerful Magnet Mount
Powerful VHB Tape Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22000424 Powerful Magnet Mount
22000399 Powerful VHB Tape Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000424 Supplier Poltex Supplier No. TXFRCUPM

Store transfer pipettes up and out of the way.

Durable and cleanable PETG construction

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Transfer Pipettes, Swabs
Material PETG
Positions 1
Sterility Non-sterile
For Use With (Equipment) Transfer pipettes, swabs
Mounting Powerful Magnet Mount
Quantity 1
Length (Metric) 102 mm
Weight (Metric) 130 g
Height (Metric) 127 mm
Width (Metric) 84 mm
Show More Show Less

consumables not included