missing translation for 'onlineSavingsMsg'
Learn More

trans-Zeatin (synthetic), 97+%

Catalog No. AC264290100
Change view
Click to view available options
Quantity:
10 mg
100 mg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC264290100 10 mg Glass bottle
AC264291000 100 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC264290100 Supplier Thermo Scientific Chemicals Supplier No. 264290100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1637-39-4 | C10H13N5O | 219.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1637-39-4
Molecular Formula C10H13N5O
Molecular Weight (g/mol) 219.25
MDL Number MFCD00213654
InChI Key UZKQTCBAMSWPJD-FARCUNLSSA-N
Synonym trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e
PubChem CID 449093
ChEBI CHEBI:16522
IUPAC Name (E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol
SMILES CC(=CCNC1=NC=NC2=C1NC=N2)CO

Specifications

Melting Point 207°C
Color Colorless to Yellow
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Merck Index 15, 10315
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and dimethylformamide
Spectrophotometry 96.0% min. (on dried substance)
Formula Weight 219.25
Percent Purity 97+%
Related Substances 2% max. (HPLC)
Physical Form Liquid
Chemical Name or Material trans-Zeatin (synthetic), 97%
Show More Show Less
RTECSNumber EM9506000

RUO – Research Use Only