accessibility menu, dialog, popup

UserName

CH3OHNHNNNHN(E)

CAS RN 1637-39-4

IUPAC Name: (2E)-2-methyl-4-[(7H-purin-6-yl)amino]but-2-en-1-ol
Synonyms: zeatin
Molecular Weight (g/mol): 219.25
Molecular Formula: C10H13N5O
InChi Key: UZKQTCBAMSWPJD-FARCUNLSSA-N
SMILES: C\C(CO)=C/CNC1=C2NC=NC2=NC=N1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (2)
Physical Form 
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)