Learn More
trans-Zeatin, 97%
Plant growth hormone that stimulates cell division | CAS: 1637-39-4 | C10H13N5O | 219.248 g/mol
Supplier: Thermo Scientific Chemicals J61972MC
Description
trans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-Zeatin regulates cell division, development, and nutrient processing. It acts as a high-affinity ligand for a number of receptors, including arabidopsis histidine kinases (AHKs), specifically AHK3, which is important in mediating plant development.
Solubility
Soluble in DMSO and DMF
Notes
Store at -20°C. Incompatible with heat and oxidizing agents.
Specifications
| trans-Zeatin | |
| 208°C to 210°C | |
| C10H13N5O | |
| 100 mg | |
| 14,10117 | |
| Soluble in DMSO and DMF | |
| CC(=CCNC1=NC=NC2=C1NC=N2)CO | |
| 219.248 | |
| CHEBI:16522 | |
| 97% |
| 1637-39-4 | |
| White | |
| MFCD00213654 | |
| 616241 | |
| trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e | |
| UZKQTCBAMSWPJD-FARCUNLSSA-N | |
| (E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol | |
| 449093 | |
| 219.25 | |
| Powder |
Chemical Identifiers
| 1637-39-4 | |
| 219.248 | |
| UZKQTCBAMSWPJD-FARCUNLSSA-N | |
| 449093 | |
| (E)-2-methyl-4-(7H-purin-6-ylamino)but-2-en-1-ol |
| C10H13N5O | |
| MFCD00213654 | |
| trans-zeatin, zeatin, e-zeatin, zeatine, e-2-methyl-4-1h-purin-6-ylamino-2-buten-1-ol, trans-zeatin synthetic, unii-7i6ooj9gr6, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, 2e, 2e-2-methyl-4-9h-purin-6-ylamino but-2-en-1-ol, 2-buten-1-ol, 2-methyl-4-1h-purin-6-ylamino-, e | |
| CHEBI:16522 | |
| CC(=CCNC1=NC=NC2=C1NC=N2)CO |
Safety and Handling
RTECSNumber : EM9506000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only