missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-4-Propyl-4'-vinylbicyclohexyl 98.0+%, TCI America™
Supplier: TCI America P231725G
Specifications
| trans,trans-4-Propyl-4′-vinylbicyclohexyl | |
| 38°C | |
| C17H30 | |
| 25 g | |
| CCCC1CCC(CC1)C2CCC(CC2)C=C | |
| 234.427 | |
| 234.43 | |
| Crystalline Lumps |
| 116020-44-1 | |
| White | |
| MFCD11045059 | |
| KHDBEDDPFRHGCN-UHFFFAOYSA-N | |
| 1-ethenyl-4-(4-propylcyclohexyl)cyclohexane | |
| 12097469 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 116020-44-1 | |
| 234.427 | |
| KHDBEDDPFRHGCN-UHFFFAOYSA-N | |
| 1-ethenyl-4-(4-propylcyclohexyl)cyclohexane |
| C17H30 | |
| MFCD11045059 | |
| 12097469 | |
| CCCC1CCC(CC1)C2CCC(CC2)C=C |
Safety and Handling
EINECSNumber : (4)-1937
TSCA : No
Recommended Storage : Refrigerator