missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-Propyl-4'-vinylbicyclohexyl 98.0+%, TCI America™

Catalog No. p-7135510
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23175G 5 g
P231725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23175G Supplier TCI America Supplier No. P23175G

Chemical Identifiers

CAS 116020-44-1
Molecular Formula C17H30
Molecular Weight (g/mol) 234.427
MDL Number MFCD11045059
InChI Key KHDBEDDPFRHGCN-UHFFFAOYSA-N
PubChem CID 12097469
IUPAC Name 1-ethenyl-4-(4-propylcyclohexyl)cyclohexane
SMILES CCCC1CCC(CC1)C2CCC(CC2)C=C

Specifications

Melting Point 38°C
Color White
Quantity 5 g
Formula Weight 234.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material trans,trans-4-Propyl-4′-vinylbicyclohexyl
EINECSNumber (4)-1937
TSCA No
Recommended Storage Refrigerator