missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-4'-Propyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 66°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 338.46 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | trans,trans-4′-Propyl-4-(3,4,5-trifluorophenyl)bicyclohexyl |
Safety and Handling
| EINECSNumber | (4)-1877 |
| TSCA | No |