missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans,trans-4'-Pentylbicyclohexyl-4-carboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 5 g |
| Formula Weight | 280.45 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | trans,trans-4′-Pentylbicyclohexyl-4-carboxylic Acid |
Safety and Handling
| TSCA | No |