accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 65355-33-1

IUPAC Name: 4'-pentyl-[1,1'-bi(cyclohexane)]-4-carboxylic acid
Synonyms: 4'-pentyl-[1,1'-bi(cyclohexane)]-4-carboxylic acid
Molecular Weight (g/mol): 280.45
Molecular Formula: C18H32O2
InChi Key: VRPANQODGRNWRV-UHFFFAOYSA-N
SMILES: CCCCCC1CCC(CC1)C1CCC(CC1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results