missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4'-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl 98.0+%, TCI America™

Catalog No. E11565G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11565G 5 g
E115625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11565G Supplier TCI America Supplier No. E11565G
Only null left

Chemical Identifiers

CAS 139215-80-8
Molecular Formula C20H27F3
Molecular Weight (g/mol) 324.431
MDL Number MFCD12911825
InChI Key DXFHMMVZUZLQFU-UHFFFAOYSA-N
Synonym 1r,4r-4-ethyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, benzene, 5-trans,trans-4'-ethyl 1,1'-bicyclohexyl-4-yl-1,2,3-trifluoro, 4-ethyl-4'-3,4,5-trifluorophenyl bi cyclohexane, trans-4-ethyl-4'-3,4,5-trifluorophenyl bicyclohexyl, 4-ethyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, 5-4-4-ethylcyclohexyl cyclohexyl-1,2,3-trifluorobenzene, acmc-20myma, 4-ethyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexyl, trans,trans-4-4'-ethylbicyclohexyl-1,2,3-trifluorobenzene
PubChem CID 14984828
IUPAC Name 5-[4-(4-ethylcyclohexyl)cyclohexyl]-1,2,3-trifluorobenzene
SMILES CCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F

Specifications

Melting Point 73°C
Color White
Quantity 5 g
Formula Weight 324.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4′-Ethyl-4-(3,4,5-trifluorophenyl)bicyclohexyl
EINECSNumber (2)-3938
TSCA No