missing translation for 'onlineSavingsMsg'
Learn More

trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl 98.0+%, TCI America™

Catalog No. p-7133341
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M26381G Supplier TCI America Supplier No. M26381G

Chemical Identifiers

CAS 155041-85-3
Molecular Formula C21H30
Molecular Weight (g/mol) 282.471
MDL Number MFCD12911573
InChI Key BCUMFFLWJNKNOU-UHFFFAOYSA-N
Synonym trans,trans-4-p-tolyl-4'-vinyl-1,1'-bi cyclohexane, trans,trans-4-p-tolyl-4'-vinyl-bicyclohexyl, 1-trans,trans-4'-ethenyl 1,1'-bicyclohexyl-4-yl-4-methylbenzene, trans,trans-4-4-methylphenyl-4'-vinylbicyclohexyl, 1-4-4-ethenylcyclohexyl cyclohexyl-4-methylbenzene, ccp-v 1, bcumfflwjnknou-ysxiemcbsa-n, 4-p-tolyl-4'-vinylbi cyclohexane, trans-4-p-tolyl-trans-4'-vinylbicyclohexyl, trans,trans-4-p-tolyl-4'-vinylbicyclohexyl
PubChem CID 20580010
IUPAC Name 1-[4-(4-ethenylcyclohexyl)cyclohexyl]-4-methylbenzene
SMILES CC1=CC=C(C=C1)C2CCC(CC2)C3CCC(CC3)C=C

Specifications

Melting Point 66°C
Color White
Quantity 1 g
Formula Weight 282.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans,trans-4-(4-Methylphenyl)-4′-vinylbicyclohexyl
EINECSNumber (4)-1940
TSCA No